Product Details
- SNP ID
-
rs13424
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:125957058 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACAAGGAGGCTAAATTTCTATCCTG[A/C]AGTTCATAAACATGTGGCGCTCGGT
- Phenotype
-
MIM: 608707
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CDON
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116746244] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDON
- Gene Name
- cell adhesion associated, oncogene regulated
- Gene
- LOC338667
- Gene Name
- V-set and immunoglobulin domain-containing protein 10-like
View Full Product Details