Product Details

SNP ID
rs1025165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125789425 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGCATAACATGCCACCTTCGCA[A/C]ACGGACAGACCGCTGTAGAAGAGGC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PATE3 PubMed Links

Gene Details

Gene
PATE3
Gene Name
prostate and testis expressed 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129883.3 3430 Missense Mutation AAA,ACA K30T NP_001123355.3
XM_017017054.1 3430 Missense Mutation AAA,ACA K18T XP_016872543.1

View Full Product Details