Product Details

SNP ID
rs1711164
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:136342669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATTTATAGGATGTAAACTTTAG[G/T]GGTTTTTTAGCCAATACAAACTGTC
Phenotype
MIM: 604358
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
STAG1 PubMed Links

Gene Details

Gene
STAG1
Gene Name
stromal antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005862.2 Intron NP_005853.2
XM_011512329.2 Intron XP_011510631.1
XM_011512331.2 Intron XP_011510633.1
XM_017005523.1 Intron XP_016861012.1
XM_017005524.1 Intron XP_016861013.1
XM_017005525.1 Intron XP_016861014.1

View Full Product Details