Product Details

SNP ID
rs1063656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176153120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGAGGGCAGCCCCCTCCCTAGAG[C/T]GGGATGGGGATGGGAGGGGGGGCGG
Phenotype
MIM: 142980 MIM: 142981 MIM: 611576
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXD3 PubMed Links

Gene Details

Gene
HOXD3
Gene Name
homeobox D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006898.4 2089 Intron NP_008829.3
XM_005246509.3 2089 Intron XP_005246566.1
XM_005246510.4 2089 Intron XP_005246567.1
XM_005246511.3 2089 Intron XP_005246568.1
XM_005246513.4 2089 Intron XP_005246570.1
XM_006712477.2 2089 Intron XP_006712540.1
XM_011511065.2 2089 Intron XP_011509367.1
XM_011511066.2 2089 Intron XP_011509368.1
Gene
HOXD4
Gene Name
homeobox D4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014621.2 2089 UTR 3 NP_055436.2
XM_005246514.4 2089 UTR 3 XP_005246571.1
Gene
MIR10B
Gene Name
microRNA 10b
There are no transcripts associated with this gene.

View Full Product Details