Product Details

SNP ID
rs1549723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:43233375 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACTTTGCAAGCCTATACCTCCTCA[A/C]CTCGGCTGAAAGCAGCCATGGACAA
Phenotype
MIM: 611800 MIM: 612053
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC01126 PubMed Links

Gene Details

Gene
LINC01126
Gene Name
long intergenic non-protein coding RNA 1126
There are no transcripts associated with this gene.

Gene
THADA
Gene Name
THADA, armadillo repeat containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083953.1 Intron NP_001077422.1
NM_001271643.1 Intron NP_001258572.1
NM_001271644.1 Intron NP_001258573.1
NM_022065.4 Intron NP_071348.3
XM_006712061.3 Intron XP_006712124.1
XM_006712062.1 Intron XP_006712125.1
XM_006712063.1 Intron XP_006712126.1
XM_006712064.2 Intron XP_006712127.1
XM_006712065.1 Intron XP_006712128.1
XM_006712066.1 Intron XP_006712129.1
XM_006712067.1 Intron XP_006712130.1
XM_006712068.3 Intron XP_006712131.1
XM_006712069.3 Intron XP_006712132.1
XM_011533015.2 Intron XP_011531317.1
XM_017004674.1 Intron XP_016860163.1
XM_017004675.1 Intron XP_016860164.1
Gene
ZFP36L2
Gene Name
ZFP36 ring finger protein like 2
There are no transcripts associated with this gene.

View Full Product Details