Product Details
- SNP ID
-
rs1028586
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:23470676 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTCTCTCAGTCCTTTTCTCCTCCC[C/T]GTGGTTCAGTAAAAGATGGTCTCTT
- Phenotype
-
MIM: 610777
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NGDN
PubMed Links
Gene Details
- Gene
- NGDN
- Gene Name
- neuroguidin
View Full Product Details