Product Details

SNP ID
rs868344
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118134404 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCACTGAGGTTCCACTTGGGGAAGA[C/T]AGCTTTGCCCATATACATCCTAGTG
Phenotype
MIM: 608256 MIM: 606565
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SCN4B PubMed Links

Gene Details

Gene
SCN4B
Gene Name
sodium voltage-gated channel beta subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142348.1 3150 UTR 3 NP_001135820.1
NM_001142349.1 3150 UTR 3 NP_001135821.1
NM_174934.3 3150 UTR 3 NP_777594.1
Gene
TMPRSS4
Gene Name
transmembrane protease, serine 4
There are no transcripts associated with this gene.

View Full Product Details