Product Details
- SNP ID
-
rs871784
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:65412835 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCTCCGGCCGACCCGAGCAGCCG[C/G]TTCCCTCCTCTCCAGGCCCCCTCCC
- Phenotype
-
MIM: 602589
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FUT8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117509929] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FUT8
- Gene Name
- fucosyltransferase 8
- Gene
- FUT8-AS1
- Gene Name
- FUT8 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details