Product Details
- SNP ID
-
rs2014123
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:92325547 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGGCAGGGGCCCTCTGAGAGTGTC[C/T]TTTACTGGAAGAACACTTAAAAGGT
- Phenotype
-
MIM: 609840
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC24A4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117243934] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC24A4
- Gene Name
- solute carrier family 24 member 4
View Full Product Details