Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_153646.3 | 270 | Silent Mutation | ACA,ACG | T58T | NP_705932.2 |
NM_153647.3 | 270 | Silent Mutation | ACA,ACG | T58T | NP_705933.2 |
NM_153648.3 | 270 | UTR 5 | NP_705934.1 | ||
XM_005267342.2 | 270 | Intron | XP_005267399.1 | ||
XM_011536436.2 | 270 | Silent Mutation | ACA,ACG | T105T | XP_011534738.1 |
XM_011536437.2 | 270 | Silent Mutation | ACA,ACG | T105T | XP_011534739.1 |
XM_011536438.2 | 270 | Silent Mutation | ACA,ACG | T105T | XP_011534740.1 |
XM_011536439.2 | 270 | Silent Mutation | ACA,ACG | T105T | XP_011534741.1 |
XM_011536440.2 | 270 | Intron | XP_011534742.1 | ||
XM_011536441.2 | 270 | Intron | XP_011534743.1 | ||
XM_011536442.2 | 270 | Intron | XP_011534744.1 |