Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001007101.2 | 2955 | UTR 3 | NP_001007102.1 | ||
NM_001261458.1 | 2955 | UTR 3 | NP_001248387.1 | ||
NM_001261459.1 | 2955 | UTR 3 | NP_001248388.1 | ||
NM_001261460.1 | 2955 | UTR 3 | NP_001248389.1 | ||
NM_031486.2 | 2955 | UTR 3 | NP_113674.1 | ||
XM_006717299.3 | 2955 | UTR 3 | XP_006717362.1 | ||
XM_011519073.2 | 2955 | UTR 3 | XP_011517375.1 | ||
XM_011519074.2 | 2955 | UTR 3 | XP_011517376.1 | ||
XM_011519075.2 | 2955 | UTR 3 | XP_011517377.1 | ||
XM_017015178.1 | 2955 | UTR 3 | XP_016870667.1 | ||
XM_017015179.1 | 2955 | UTR 3 | XP_016870668.1 | ||
XM_017015180.1 | 2955 | UTR 3 | XP_016870669.1 | ||
XM_017015181.1 | 2955 | UTR 3 | XP_016870670.1 |