Product Details

SNP ID
rs7182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73712097 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGAGTAATAGCTAATAAATGCACA[C/T]TTCCTTCCATTAATATAAATCTGGA
Phenotype
MIM: 610062 MIM: 604010
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAL1 PubMed Links

Gene Details

Gene
DNAL1
Gene Name
dynein axonemal light chain 1
There are no transcripts associated with this gene.

Gene
ELMSAN1
Gene Name
ELM2 and Myb/SANT domain containing 1
There are no transcripts associated with this gene.

Gene
PNMA1
Gene Name
paraneoplastic Ma antigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006029.4 2329 UTR 3 NP_006020.4

View Full Product Details