Product Details

SNP ID
rs707560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:59152257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTAAATCCACTACAATGTAAGA[C/T]AGGCATAAATTCACCTAAAAGTGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF831 PubMed Links

Gene Details

Gene
ZNF831
Gene Name
zinc finger protein 831
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178457.2 Intron NP_848552.1
XM_005260272.3 Intron XP_005260329.1
XM_005260273.3 Intron XP_005260330.1
XM_006723698.3 Intron XP_006723761.1
XM_011528534.2 Intron XP_011526836.1
XM_011528536.2 Intron XP_011526838.1
XM_011528537.2 Intron XP_011526839.1
XM_011528538.2 Intron XP_011526840.1
XM_011528540.2 Intron XP_011526842.1
XM_017027642.1 Intron XP_016883131.1
XM_017027643.1 Intron XP_016883132.1
XM_017027644.1 Intron XP_016883133.1

View Full Product Details