Product Details

SNP ID
rs1061093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62567436 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATCTTCTCACACAGTTTCACTTC[C/G]CCCAAGACAGCCCGGAACTGGGGCT
Phenotype
MIM: 130593 MIM: 610641
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EEF1G PubMed Links

Gene Details

Gene
EEF1G
Gene Name
eukaryotic translation elongation factor 1 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001404.4 Intron NP_001395.1
Gene
MIR6747
Gene Name
microRNA 6747
There are no transcripts associated with this gene.

Gene
TUT1
Gene Name
terminal uridylyl transferase 1, U6 snRNA-specific
There are no transcripts associated with this gene.

View Full Product Details