Product Details

SNP ID
rs720669
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103497750 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATATTGAATTCAACTAATATCACA[A/G]AGAGATGTTCTTAATCTTCTAGACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB40A PubMed Links

Gene Details

Gene
RAB40A
Gene Name
RAB40A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080879.2 3823 Intron NP_543155.2
XM_005262083.4 3823 UTR 3 XP_005262140.1
XM_011530872.2 3823 UTR 3 XP_011529174.1

View Full Product Details