Product Details

SNP ID
rs1052428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103711037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACAGGCAACTTAGCTGGTGAAAG[G/T]ACTCTGGATTGGTTGGCAGTCTGCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GLRA4 PubMed Links

Gene Details

Gene
GLRA4
Gene Name
glycine receptor alpha 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024452.2 129 Intron NP_001019623.2
NM_001172285.1 129 Intron NP_001165756.1
Gene
TMEM31
Gene Name
transmembrane protein 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182541.2 129 UTR 5 NP_872347.2

View Full Product Details