Product Details
- SNP ID
-
rs2316872
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:2907483 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GCACTTCATAATGTCATAGGAATTA[C/T]TTTTCCCCCATCCTACACTTCAGTA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF57
PubMed Links
Gene Details
- Gene
- ZNF57
- Gene Name
- zinc finger protein 57
View Full Product Details