Product Details

SNP ID
rs2116932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39446053 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTGTGGGAGGCTCGAGGGGTTCA[C/T]AGCGGGCTCTGGCTTCTGGGAACTC
Phenotype
MIM: 602102
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC723805 PubMed Links

Gene Details

Gene
LOC723805
Gene Name
interleukin-like
There are no transcripts associated with this gene.

Gene
SUPT5H
Gene Name
SPT5 homolog, DSIF elongation factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111020.2 Intron NP_001104490.1
NM_001130824.1 Intron NP_001124296.1
NM_001130825.1 Intron NP_001124297.1
NM_001319990.1 Intron NP_001306919.1
NM_001319991.1 Intron NP_001306920.1
NM_003169.3 Intron NP_003160.2
XM_017027174.1 Intron XP_016882663.1

View Full Product Details