Product Details

SNP ID
rs7996068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:42565254 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTTTTTTCTTTAGTTTATCTGGT[A/G]GCTTTGTCTAGTTGTTATGGTTATG
Phenotype
MIM: 602642
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNFSF11 PubMed Links

Gene Details

Gene
TNFSF11
Gene Name
tumor necrosis factor superfamily member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003701.3 Intron NP_003692.1
NM_033012.3 Intron NP_143026.1
XM_011535280.2 Intron XP_011533582.1
XM_017020802.1 Intron XP_016876291.1
XM_017020803.1 Intron XP_016876292.1

View Full Product Details