Product Details
- SNP ID
-
rs2015209
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:48204408 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTTTTCAAGGACAGGGAGTGTTTTG[A/G]TGAATTCCTATGTAATAAGGAGATT
- Phenotype
-
MIM: 609051
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C19orf68
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115132442] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C19orf68
- Gene Name
- chromosome 19 open reading frame 68
There are no transcripts associated with this gene.
- Gene
- CARD8
- Gene Name
- caspase recruitment domain family member 8
View Full Product Details