Product Details

SNP ID
rs8021195
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.14:23017877 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTATTCATGTCTGCTAGACGTCTCT[A/G]TTGTAGAAGTATATTTGCCATTTGT
Phenotype
MIM: 600306
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf93 PubMed Links

Gene Details

Gene
C14orf93
Gene Name
chromosome 14 open reading frame 93
There are no transcripts associated with this gene.

Gene
PSMB5
Gene Name
proteasome subunit beta 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130725.1 Intron NP_001124197.1
NM_001144932.2 Intron NP_001138404.1
NM_002797.4 Intron NP_002788.1
XM_005267871.2 Intron XP_005267928.1

View Full Product Details