Product Details
- SNP ID
-
rs2231297
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:52109736 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCAGGACTCCTTGTATGCATTCAG[A/G]TAAAAACGTCTGGGGTGGGGGAAGG
- Phenotype
-
MIM: 606910
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BCL2L10
PubMed Links
Gene Details
- Gene
- BCL2L10
- Gene Name
- BCL2 like 10
View Full Product Details