Product Details

SNP ID
rs1128154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121631769 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACCTCATCTGGTTAGACATTTGCA[G/T]CAGGAATAGGGAGGGGGTGGAGGCA
Phenotype
MIM: 609107 MIM: 601306
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXO40 PubMed Links

Gene Details

Gene
FBXO40
Gene Name
F-box protein 40
There are no transcripts associated with this gene.

Gene
HCLS1
Gene Name
hematopoietic cell-specific Lyn substrate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292041.1 1556 UTR 3 NP_001278970.1
NM_005335.5 1556 UTR 3 NP_005326.2

View Full Product Details