Product Details
- SNP ID
-
rs1210015
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:30720868 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTCTGGGTGGCCGGAGCCGGAGCA[C/G]AGCCTGGGTTTGGAGTGAGTGCCTG
- Phenotype
-
MIM: 602988
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PCDH7
PubMed Links
Gene Details
- Gene
- PCDH7
- Gene Name
- protocadherin 7
View Full Product Details