Product Details

SNP ID
rs1634324
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:13044597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCAGAGGCTGGCACCGCGAAG[A/C]TCCTCCGAGGGCCTGGGTCCTCTGT
Phenotype
MIM: 300479
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM9C PubMed Links

Gene Details

Gene
FAM9C
Gene Name
family with sequence similarity 9 member C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174901.5 202 UTR 5 NP_777561.1
XM_005274460.2 202 Intron XP_005274517.1
XM_017029321.1 202 Intron XP_016884810.1
Gene
LOC105373133
Gene Name
uncharacterized LOC105373133
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011545614.2 202 Missense Mutation GAT,GCT D15A XP_011543916.1

View Full Product Details