Product Details

SNP ID
rs1701704
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56018703 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCAGTGTCCACTAGAACTTGTCA[G/T]GCAGGAATGTGTATCTAGTGTGGCC
Phenotype
MIM: 606239
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IKZF4 PubMed Links

Gene Details

Gene
IKZF4
Gene Name
IKAROS family zinc finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022465.3 Intron NP_071910.3
XM_005269086.3 Intron XP_005269143.1
XM_005269089.2 Intron XP_005269146.1
XM_005269090.3 Intron XP_005269147.1
XM_005269093.2 Intron XP_005269150.1
XM_011538664.1 Intron XP_011536966.1
XM_011538666.2 Intron XP_011536968.1
XM_011538669.2 Intron XP_011536971.1
XM_017019805.1 Intron XP_016875294.1
XM_017019806.1 Intron XP_016875295.1
XM_017019807.1 Intron XP_016875296.1
XM_017019808.1 Intron XP_016875297.1
XM_017019809.1 Intron XP_016875298.1
XM_017019810.1 Intron XP_016875299.1
XM_017019811.1 Intron XP_016875300.1
XM_017019812.1 Intron XP_016875301.1
XM_017019813.1 Intron XP_016875302.1
XM_017019814.1 Intron XP_016875303.1
XM_017019815.1 Intron XP_016875304.1
XM_017019816.1 Intron XP_016875305.1
Gene
LOC105369781
Gene Name
uncharacterized LOC105369781
There are no transcripts associated with this gene.

View Full Product Details