Product Details

SNP ID
rs1001242
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:149452585 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TACTGCCCCCACACAAGGCCCTGAA[C/G]AGGGCCTGCGAAGCACTGGGCCCTA
Phenotype
MIM: 611106
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZC3H12D PubMed Links

Gene Details

Gene
ZC3H12D
Gene Name
zinc finger CCCH-type containing 12D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207360.2 Intron NP_997243.2

View Full Product Details