Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001318336.1 | 3988 | UTR 3 | NP_001305265.1 | ||
NM_001318337.1 | 3988 | UTR 3 | NP_001305266.1 | ||
NM_001318338.1 | 3988 | UTR 3 | NP_001305267.1 | ||
NM_018027.4 | 3988 | UTR 3 | NP_060497.3 | ||
XM_005252489.2 | 3988 | UTR 3 | XP_005252546.1 | ||
XM_005252490.2 | 3988 | UTR 3 | XP_005252547.1 | ||
XM_005252495.4 | 3988 | UTR 3 | XP_005252552.1 | ||
XM_006717457.2 | 3988 | UTR 3 | XP_006717520.1 | ||
XM_006717458.3 | 3988 | UTR 3 | XP_006717521.1 | ||
XM_006717459.2 | 3988 | UTR 3 | XP_006717522.1 | ||
XM_006717460.2 | 3988 | UTR 3 | XP_006717523.1 | ||
XM_006717464.1 | 3988 | UTR 3 | XP_006717527.1 | ||
XM_011519539.1 | 3988 | UTR 3 | XP_011517841.1 | ||
XM_011519540.1 | 3988 | UTR 3 | XP_011517842.1 | ||
XM_011519541.1 | 3988 | UTR 3 | XP_011517843.1 | ||
XM_011519543.2 | 3988 | UTR 3 | XP_011517845.1 | ||
XM_017016393.1 | 3988 | UTR 3 | XP_016871882.1 | ||
XM_017016394.1 | 3988 | UTR 3 | XP_016871883.1 | ||
XM_017016395.1 | 3988 | UTR 3 | XP_016871884.1 | ||
XM_017016396.1 | 3988 | UTR 3 | XP_016871885.1 |