Product Details
- SNP ID
-
rs1453162
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:206943008 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTTAGGAAGTAACAGACTCACATT[A/T]TGTAGACATTGTAGCCAAGAGTAAG
- Phenotype
-
MIM: 609563
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CPO
PubMed Links
Gene Details
- Gene
- CPO
- Gene Name
- carboxypeptidase O
View Full Product Details