Product Details

SNP ID
rs6643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:85597128 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACAAATTCCTGTCCCTTGTGCCGC[C/T]ATGAGCTGCCCACTGATGACGACAC
Phenotype
MIM: 612490 MIM: 616757 MIM: 611594 MIM: 607029
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2orf68 PubMed Links

Gene Details

Gene
C2orf68
Gene Name
chromosome 2 open reading frame 68
There are no transcripts associated with this gene.

Gene
RNF181
Gene Name
ring finger protein 181
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016494.3 402 Missense Mutation CAT,TAT H118Y NP_057578.1
XM_005264359.3 402 Silent Mutation GCC,GCT A131A XP_005264416.1
Gene
TMEM150A
Gene Name
transmembrane protein 150A
There are no transcripts associated with this gene.

Gene
USP39
Gene Name
ubiquitin specific peptidase 39
There are no transcripts associated with this gene.

Gene
VAMP5
Gene Name
vesicle associated membrane protein 5
There are no transcripts associated with this gene.

View Full Product Details