Product Details

SNP ID
rs1168773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:66196952 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTGGCATGTGGTTGTATGCAAAT[A/G]CAAAGATTCCAAATACGGATGTGTT
Phenotype
MIM: 604459
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRAK3 PubMed Links

Gene Details

Gene
IRAK3
Gene Name
interleukin 1 receptor associated kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142523.1 Intron NP_001135995.1
NM_007199.2 Intron NP_009130.2

View Full Product Details