Product Details

SNP ID
rs4832
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159778994 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTTAATGTGTAATACTCAACTCA[A/C]GGTACAAGACAATTGCATTTAACAT
Phenotype
MIM: 100678 MIM: 186980
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
ACAT2 PubMed Links

Gene Details

Gene
ACAT2
Gene Name
acetyl-CoA acetyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303253.1 1957 UTR 3 NP_001290182.1
NM_005891.2 1957 UTR 3 NP_005882.2
Gene
SNORA20
Gene Name
small nucleolar RNA, H/ACA box 20
There are no transcripts associated with this gene.

Gene
SNORA29
Gene Name
small nucleolar RNA, H/ACA box 29
There are no transcripts associated with this gene.

Gene
TCP1
Gene Name
t-complex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008897.1 1957 UTR 3 NP_001008897.1
NM_030752.2 1957 UTR 3 NP_110379.2

View Full Product Details