Product Details

SNP ID
rs1048210
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:109440889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGCCTGGGCGCCCGGATTTCGG[C/T]AGCGGATCGCCTTTCCGGGTTGGCG
Phenotype
MIM: 607129 MIM: 603498
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MICAL1 PubMed Links
Additional Information
For this assay, SNP(s) [rs115963563] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MICAL1
Gene Name
microtubule associated monooxygenase, calponin and LIM domain containing 1
There are no transcripts associated with this gene.

Gene
PPIL6
Gene Name
peptidylprolyl isomerase like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111298.2 283 UTR 5 NP_001104768.2
NM_001286360.1 283 UTR 5 NP_001273289.1
NM_001286361.1 283 UTR 5 NP_001273290.1
NM_173672.4 283 UTR 5 NP_775943.1
XM_011535765.2 283 Intron XP_011534067.1
XM_011535766.2 283 Intron XP_011534068.1
XM_011535767.2 283 Intron XP_011534069.1
XM_011535769.2 283 Intron XP_011534071.1
XM_017010774.1 283 Intron XP_016866263.1
Gene
SMPD2
Gene Name
sphingomyelin phosphodiesterase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003080.2 283 UTR 5 NP_003071.2
XM_005267109.1 283 Intron XP_005267166.1
XM_011536079.1 283 Intron XP_011534381.1

View Full Product Details