Product Details
- SNP ID
-
rs707304
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:52401569 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCTACTGCAACTACACAGCCCCC[C/T]TTCCTTTTCTGAGAGCGTTTTGTAT
- Phenotype
-
MIM: 615580
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF528
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114764188] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ZNF528
- Gene Name
- zinc finger protein 528
- Gene
- ZNF528-AS1
- Gene Name
- ZNF528 antisense RNA 1
There are no transcripts associated with this gene.
- Gene
- ZNF880
- Gene Name
- zinc finger protein 880
There are no transcripts associated with this gene.
View Full Product Details