Product Details

SNP ID
rs1042487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:48158885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTTTGCTTAAGTTTCTCTGCAG[C/G]AAAGAGGGTAAAGGGGGATGGAGGG
Phenotype
MIM: 116947
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CDC25A PubMed Links

Gene Details

Gene
CDC25A
Gene Name
cell division cycle 25A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001789.2 2091 UTR 3 NP_001780.2
NM_201567.1 2091 UTR 3 NP_963861.1
XM_006713434.2 2091 UTR 3 XP_006713497.1
XM_006713435.2 2091 UTR 3 XP_006713498.1
XM_011534316.1 2091 UTR 3 XP_011532618.1

View Full Product Details