Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001130413.3 | Intron | NP_001123885.2 | |||
XM_011541899.2 | Intron | XP_011540201.1 | |||
XM_011541901.2 | Intron | XP_011540203.1 | |||
XM_011541902.2 | Intron | XP_011540204.1 | |||
XM_011541905.2 | Intron | XP_011540207.1 | |||
XM_011541906.2 | Intron | XP_011540208.1 | |||
XM_011541908.2 | Intron | XP_011540210.1 | |||
XM_011541920.2 | Intron | XP_011540222.1 | |||
XM_011541925.2 | Intron | XP_011540227.1 | |||
XM_011541929.2 | Intron | XP_011540231.1 | |||
XM_011541932.2 | Intron | XP_011540234.1 | |||
XM_011541933.2 | Intron | XP_011540235.1 | |||
XM_017002037.1 | Intron | XP_016857526.1 | |||
XM_017002038.1 | Intron | XP_016857527.1 | |||
XM_017002039.1 | Intron | XP_016857528.1 | |||
XM_017002040.1 | Intron | XP_016857529.1 | |||
XM_017002041.1 | Intron | XP_016857530.1 | |||
XM_017002042.1 | Intron | XP_016857531.1 | |||
XM_017002043.1 | Intron | XP_016857532.1 | |||
XM_017002044.1 | Intron | XP_016857533.1 |