Product Details

SNP ID
rs1044885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197392203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTAGCTCTTCCTCTATGACCAG[C/T]TCTACACTGATCTGCAATGTTTAAA
Phenotype
MIM: 605590
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SF3B1 PubMed Links

Gene Details

Gene
SF3B1
Gene Name
splicing factor 3b subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005526.2 4109 Intron NP_001005526.1
NM_001308824.1 4109 Intron NP_001295753.1
NM_012433.3 4109 UTR 3 NP_036565.2

View Full Product Details