Product Details

SNP ID
hCV8752279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3189071 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAAAAAGCGAAACTCCTGACACC[C/T]TTAAAACAGAAAACATTGTTATTCA
Phenotype
MIM: 608998
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TSSC1 PubMed Links

Gene Details

Gene
TSSC1
Gene Name
tumor suppressing subtransferable candidate 1
There are no transcripts associated with this gene.

View Full Product Details