Product Details

SNP ID
rs8052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:150542155 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGACAACTTGTTGAAATGTTAGAG[C/T]TAAACCACAAAGAGTAACTGAGTAA
Phenotype
MIM: 609234
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF2A PubMed Links

Gene Details

Gene
EIF2A
Gene Name
eukaryotic translation initiation factor 2A
There are no transcripts associated with this gene.

Gene
SERP1
Gene Name
stress-associated endoplasmic reticulum protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014445.3 3675 UTR 3 NP_055260.1
XM_011512698.2 3675 UTR 3 XP_011511000.1

View Full Product Details