Product Details

SNP ID
rs1047296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30156360 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTACCCAGGCGGTGGTGTCGAT[C/T]TACGTTCCAATTGGGGCCGTACCAT
Phenotype
MIM: 612963 MIM: 611940
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCTN6 PubMed Links

Gene Details

Gene
DCTN6
Gene Name
dynactin subunit 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006571.3 64 UTR 5 NP_006562.1
Gene
MBOAT4
Gene Name
membrane bound O-acyltransferase domain containing 4
There are no transcripts associated with this gene.

View Full Product Details