Product Details

SNP ID
rs1677235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:55544858 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGTTTTGAATCTTAATGAGCACA[A/C]CTCCAACTGTCTCCATTCTTGATCA
Phenotype
MIM: 606902
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PYGO1 PubMed Links

Gene Details

Gene
PYGO1
Gene Name
pygopus family PHD finger 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015617.2 2820 UTR 3 NP_056432.1
XM_005254300.4 2820 UTR 3 XP_005254357.1
XM_011521446.2 2820 UTR 3 XP_011519748.1

View Full Product Details