Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001306153.1 | 411 | Intron | NP_001293082.1 | ||
NM_001306154.1 | 411 | UTR 5 | NP_001293083.1 | ||
NM_001306155.1 | 411 | Intron | NP_001293084.1 | ||
NM_001306156.1 | 411 | Intron | NP_001293085.1 | ||
NM_012073.4 | 411 | UTR 5 | NP_036205.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001258388.1 | 411 | Intron | NP_001245317.1 | ||
NM_001258389.1 | 411 | Intron | NP_001245318.1 | ||
NM_199133.3 | 411 | Intron | NP_954584.2 | ||
XM_011513964.1 | 411 | Intron | XP_011512266.1 |