Product Details

SNP ID
rs1052457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50702672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTCATATTCCAAAGGAATAGAA[A/C]AGCTGTTTGCAATACTTTTTATGCC
Phenotype
MIM: 608238
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPPL2A PubMed Links

Gene Details

Gene
SPPL2A
Gene Name
signal peptide peptidase like 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032802.3 6924 Intron NP_116191.2
XM_005254722.3 6924 UTR 3 XP_005254779.1
XM_017022680.1 6924 Intron XP_016878169.1
XM_017022681.1 6924 Intron XP_016878170.1

View Full Product Details