Product Details

SNP ID
rs1626437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42161080 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTATTTTTTTATTATGCAAAACACA[A/C]GGCATATTAAAGATGGGCAGTGCAC
Phenotype
MIM: 612188
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PLA2G4F PubMed Links

Gene Details

Gene
PLA2G4F
Gene Name
phospholipase A2 group IVF
There are no transcripts associated with this gene.

Gene
VPS39
Gene Name
VPS39, HOPS complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301138.1 3241 Intron NP_001288067.1
NM_015289.3 3241 Intron NP_056104.2
XM_011521403.2 3241 Intron XP_011519705.1
XM_011521404.2 3241 Intron XP_011519706.1
XM_017022036.1 3241 UTR 3 XP_016877525.1
XM_017022037.1 3241 UTR 3 XP_016877526.1
XM_017022038.1 3241 Intron XP_016877527.1
XM_017022039.1 3241 Intron XP_016877528.1

View Full Product Details