Product Details

SNP ID
rs1015600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:36841129 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGATTAGTAGATGTGTATTCCTA[A/C]GTATTCTACAGCAATGAGAAATAAG
Phenotype
MIM: 614765
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
STRN PubMed Links

Gene Details

Gene
STRN
Gene Name
striatin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003162.3 10679 UTR 3 NP_003153.2
XM_005264519.4 10679 UTR 3 XP_005264576.1
XM_011533073.2 10679 UTR 3 XP_011531375.1

View Full Product Details