Product Details
- SNP ID
-
rs1802643
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:177332165 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACGGCGCAGACAACGATGCCCAGG[C/A]GAGCGCACAGCAGCAGCAGGAACAC
- Phenotype
-
MIM: 609551
- Polymorphism
- C/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
LMAN2
PubMed Links
Gene Details
- Gene
- LMAN2
- Gene Name
- lectin, mannose binding 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006816.2 |
1229 |
Missense Mutation |
CGC,CTC |
R331L |
NP_006807.1 |
View Full Product Details