Product Details

SNP ID
rs1048238
Assay Type
Functionally Tested
NCBI dbSNP Submissions
43
Location
Chr.1:16015154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTGCCTGTGACCTTCCCTCTGTC[C/T]CTCCCACTCCCCTTGGCCAAGAGGG
Phenotype
MIM: 602024 MIM: 610692
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C1orf64 PubMed Links
Additional Information
For this assay, SNP(s) [rs77577644] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C1orf64
Gene Name
chromosome 1 open reading frame 64
There are no transcripts associated with this gene.

Gene
CLCNKA
Gene Name
chloride voltage-gated channel Ka
There are no transcripts associated with this gene.

Gene
HSPB7
Gene Name
heat shock protein family B (small) member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014424.4 1766 UTR 3 NP_055239.1
XM_011541249.1 1766 UTR 3 XP_011539551.1
XM_011541250.2 1766 UTR 3 XP_011539552.1

View Full Product Details