Product Details

SNP ID
rs945403
Assay Type
Functionally Tested
NCBI dbSNP Submissions
71
Location
Chr.1:16047835 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCGAGATCGTAATGTGAAAACATC[A/G]CACAGGTAGAGTGTTGAGATTTTTA
Phenotype
MIM: 602023
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CLCNKB PubMed Links

Gene Details

Gene
CLCNKB
Gene Name
chloride voltage-gated channel Kb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000085.4 Intron NP_000076.2
NM_001165945.2 Intron NP_001159417.2
Gene
FAM131C
Gene Name
family with sequence similarity 131 member C
There are no transcripts associated with this gene.

View Full Product Details