Product Details

SNP ID
rs1749955
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:228408549 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACACACCCAGGGCCCACAACGC[A/G]TCCCCGGTGATGTTCATGCCCACCT
Phenotype
MIM: 607868 MIM: 606123
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM11 PubMed Links

Gene Details

Gene
TRIM11
Gene Name
tripartite motif containing 11
There are no transcripts associated with this gene.

Gene
TRIM17
Gene Name
tripartite motif containing 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024940.2 2003 Silent Mutation GAC,GAT D362D NP_001020111.1
NM_001134855.1 2003 Intron NP_001128327.1
NM_016102.3 2003 Silent Mutation GAC,GAT D362D NP_057186.1
XM_006711779.2 2003 Silent Mutation GAC,GAT D362D XP_006711842.1
XM_011544209.2 2003 Silent Mutation GAC,GAT D362D XP_011542511.1
XM_011544210.2 2003 Silent Mutation GAC,GAT D362D XP_011542512.1
XM_011544211.2 2003 Silent Mutation GAC,GAT D335D XP_011542513.1
XM_017001418.1 2003 UTR 3 XP_016856907.1
XM_017001419.1 2003 UTR 3 XP_016856908.1

View Full Product Details