Product Details

SNP ID
rs824402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:83954943 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGATGGAGCAGGTTGGCTCACT[C/T]GTTTGGTGAAGGGGCTGGGGTGCCT
Phenotype
MIM: 607975
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105371371 PubMed Links

Gene Details

Gene
LOC105371371
Gene Name
uncharacterized LOC105371371
There are no transcripts associated with this gene.

Gene
OSGIN1
Gene Name
oxidative stress induced growth inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182981.2 Intron NP_892026.1

View Full Product Details